siRNA Details
| Gene Symbol: LOC100499194 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034130.1 | 1 | 400 | |||
| GenBank mRNA | BG772716.1 | - | 2 | 208 | ||
| DB024523.1 | - | 431 | ||||
| Gene Symbol: LINC01191 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034128.1 | 1 | 13 | |||

LOC100499194
Genomic Map