siRNA Details
| Gene Symbol: LINC01297 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_132385.1 | 3 | 463 | |||
| GenBank mRNA | AI023613.1 | - | 1 | 108 | ||
| BF001177.1 | - | 1 | 117 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01297| Gene Symbol: LINC01297 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_132385.1 | 3 | 463 | |||
| GenBank mRNA | AI023613.1 | - | 1 | 108 | ||
| BF001177.1 | - | 1 | 117 | |||