siRNA Details
Gene Symbol: THUMPD3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK128398.1 | - | 1 | 202 | ||
BC132680.1 | - | 1 | 176 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: THUMPD3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK128398.1 | - | 1 | 202 | ||
BC132680.1 | - | 1 | 176 |