siRNA Details
| Gene Symbol: ZNF654 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017006789.1 | XP_016862278.1 | 4 | 620 | ||
| XM_017006790.1 | XP_016862279.1 | 4 | 632 | |||
| XM_017006794.1 | XP_016862283.1 | 2 | 197 | |||
| XM_017006795.1 | XP_016862284.1 | 4 | 628 | |||

ZNF654
Genomic Map