siRNA Details
| Gene Symbol: LINC00969 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK128346.1 | - | 1 | 1897 | ||
| BC144457.1 | - | 1 | 1424 | |||
| DA875776.1 | - | 3 | 518 | |||
| KJ535002.1 | - | 1 | 14 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00969| Gene Symbol: LINC00969 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK128346.1 | - | 1 | 1897 | ||
| BC144457.1 | - | 1 | 1424 | |||
| DA875776.1 | - | 3 | 518 | |||
| KJ535002.1 | - | 1 | 14 | |||