siRNA Details
| Gene Symbol: DANCR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024031.1 | 3 | 354 | |||
| GenBank mRNA | BC015222.1 | - | 2 | 292 | ||
| BC045566.1 | - | 3 | 303 | |||
| D28589.1 | - | 2 | 291 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DANCR| Gene Symbol: DANCR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024031.1 | 3 | 354 | |||
| GenBank mRNA | BC015222.1 | - | 2 | 292 | ||
| BC045566.1 | - | 3 | 303 | |||
| D28589.1 | - | 2 | 291 | |||