siRNA Details
| Gene Symbol: THEGL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256475.1 | NP_001243404.1 | 8 | 1282 | ||
| XM_011534357.1 | XP_011532659.1 | 8 | 1316 | |||
| XM_011534361.2 | XP_011532663.1 | 8 | 1316 | |||
| XM_011534363.2 | XP_011532665.1 | 8 | 1316 | |||
| XM_011534364.1 | XP_011532666.1 | 8 | 1316 | |||
| GenBank mRNA | DY655427.1 | - | 3 | 274 | ||

THEGL
Genomic Map