siRNA Details
| Gene Symbol: RUFY3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037442.3 | NP_001032519.1 | 18 | 2483 | ||
| NM_001291993.1 | NP_001278922.1 | 19 | 1875 | |||
| XM_005265656.2 | XP_005265713.1 | 19 | 2521 | |||
| XM_005265658.1 | XP_005265715.1 | 18 | 1797 | |||
| XM_011531750.2 | XP_011530052.1 | 19 | 2119 | |||
| XM_011531751.2 | XP_011530053.1 | 17 | 2063 | |||
| XM_011531752.2 | XP_011530054.1 | 17 | 2104 | |||
| XM_011531754.2 | XP_011530056.1 | 16 | 1998 | |||
| GenBank mRNA | DC299541.1 | - | 1 | 41 | ||

RUFY3
Genomic Map