siRNA Details
| Gene Symbol: TMEM150C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080506.1 | NP_001073975.1 | 2, 3 | 146 | ||
| GenBank mRNA | AK297434.1 | - | 2, 3 | 273 | ||
| AK297512.1 | - | 2, 3 | 151 | |||
| AK299942.1 | - | 2, 3 | 136 | |||
| AK301116.1 | - | 2, 3 | 120 | |||
| BC147019.1 | - | 2, 3 | 129 | 9056947 | ||
| BC147027.1 | - | 2, 3 | 129 | 9056955 | ||

TMEM150C
Genomic Map