siRNA Details
| Gene Symbol: KLHL8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001292003.1 | NP_001278932.1 | 1 | 118 | ||
| GenBank mRNA | AK128827.1 | - | 1 | 118 | ||
| HY117925.1 | - | 1 | 225 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KLHL8| Gene Symbol: KLHL8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001292003.1 | NP_001278932.1 | 1 | 118 | ||
| GenBank mRNA | AK128827.1 | - | 1 | 118 | ||
| HY117925.1 | - | 1 | 225 | |||