siRNA Details
| Gene Symbol: LOC100507639 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121625.1 | 3 | 660 | |||
| GenBank mRNA | AI521827.1 | - | 2 | 362 | ||
| AK124399.1 | - | 1 | 2479 | |||
| BI762316.1 | - | 3 | 648 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100507639| Gene Symbol: LOC100507639 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121625.1 | 3 | 660 | |||
| GenBank mRNA | AI521827.1 | - | 2 | 362 | ||
| AK124399.1 | - | 1 | 2479 | |||
| BI762316.1 | - | 3 | 648 | |||