siRNA Details
| Gene Symbol: LOC441052 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134656.1 | 1 | 494 | |||
| GenBank mRNA | AF131741.1 | - | 1 | 494 | ||
| Gene Symbol: GALNTL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017008243.1 | XP_016863732.1 | 1 | 421 | ||
| GenBank mRNA | AK124240.1 | - | 1 | 857 | ||

LOC441052
Genomic Map