siRNA Details
| Gene Symbol: GALNTL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017008243.1 | XP_016863732.1 | 1 | 67 | ||
| GenBank mRNA | AK124240.1 | - | 1 | 503 | ||
| Gene Symbol: LOC441052 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134656.1 | 1 | 848 | |||
| GenBank mRNA | AF131741.1 | - | 1 | 848 | ||
| DA181879.1 | - | 1 | 101 | |||

GALNTL6
Genomic Map