siRNA Details
| Gene Symbol: LINC01262 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121679.1 | 2 | 310 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01262| Gene Symbol: LINC01262 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121679.1 | 2 | 310 | |||