siRNA Details
| Gene Symbol: MGC39584 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038377.1 | 3 | 549 | |||
| GenBank mRNA | BC029568.1 | - | 3 | 549 | ||
| Gene Symbol: LOC102723847 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC033389.1 | - | 2 | 358 | ||

MGC39584
Genomic Map