siRNA Details
| Gene Symbol: GUSBP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_132999.1 | 5 | 4762 | |||
| GenBank mRNA | AK026691.1 | - | 1 | 2305 | ||
| AL831827.1 | - | 1 | 5142 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GUSBP4| Gene Symbol: GUSBP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_132999.1 | 5 | 4762 | |||
| GenBank mRNA | AK026691.1 | - | 1 | 2305 | ||
| AL831827.1 | - | 1 | 5142 | |||