siRNA Details
| Gene Symbol: JMY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152405.4 | NP_689618.4 | 1 | 1096 | ||
| XM_005248430.2 | XP_005248487.1 | 1 | 1096 | |||
| XM_011543155.2 | XP_011541457.1 | 1 | 1096 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

JMY| Gene Symbol: JMY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152405.4 | NP_689618.4 | 1 | 1096 | ||
| XM_005248430.2 | XP_005248487.1 | 1 | 1096 | |||
| XM_011543155.2 | XP_011541457.1 | 1 | 1096 | |||