siRNA Details
| Gene Symbol: NR2F1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109825.1 | 1 | 818 | |||
| GenBank mRNA | AK124699.1 | - | 1 | 268 | ||
| BC044619.1 | - | 1 | 294 | |||
| BC048999.1 | - | 1 | 294 | |||
| BC144330.1 | - | 1 | 818 | |||
| BC144331.1 | - | 1 | 818 | |||

NR2F1-AS1
Genomic Map