siRNA Details
| Gene Symbol: LOC100652833 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011543783.2 | XP_011542085.1 | 1 | 1653 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100652833| Gene Symbol: LOC100652833 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011543783.2 | XP_011542085.1 | 1 | 1653 | ||