siRNA Details
Gene Symbol: LOC100652833 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011543783.2 | XP_011542085.1 | 1 | 770 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100652833 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011543783.2 | XP_011542085.1 | 1 | 770 |