siRNA Details
| Gene Symbol: LOC101927421 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BU517015.1 | - | 229 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927421| Gene Symbol: LOC101927421 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BU517015.1 | - | 229 | |||