siRNA Details
| Gene Symbol: CCDC192 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317938.1 | NP_001304867.1 | 3 | 209 | ||
| XM_017009805.1 | XP_016865294.1 | 2 | 271 | |||
| XM_017009806.1 | XP_016865295.1 | 3 | 175 | |||
| XM_017009807.1 | XP_016865296.1 | 3 | 166 | |||
| XM_017009808.1 | XP_016865297.1 | 2 | 97 | |||
| XM_017009809.1 | XP_016865298.1 | 3 | 240 | |||
| XM_017009810.1 | XP_016865299.1 | 3 | 239 | |||
| XM_017009811.1 | XP_016865300.1 | 3 | 239 | |||
| XM_017009812.1 | XP_016865301.1 | 3 | 240 | |||
| XM_017009813.1 | XP_016865302.1 | 3 | 238 | |||
| GenBank mRNA | HY052603.1 | - | 3 | 177 | ||

CCDC192
Genomic Map