siRNA Details
| Gene Symbol: MEIKIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303622.1 | NP_001290551.1 | 8, 9 | 903 | ||
| GenBank mRNA | AB987829.1 | - | 8, 9 | 710 | ||
| DY654845.1 | - | 4, 5 | 302 | |||
| HY202839.1 | - | 4, 5 | 436 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MEIKIN| Gene Symbol: MEIKIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303622.1 | NP_001290551.1 | 8, 9 | 903 | ||
| GenBank mRNA | AB987829.1 | - | 8, 9 | 710 | ||
| DY654845.1 | - | 4, 5 | 302 | |||
| HY202839.1 | - | 4, 5 | 436 | |||