siRNA Details
| Gene Symbol: ZNF391 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001076781.2 | NP_001070249.1 | 1 | 229 | ||
| NM_001322289.1 | NP_001309218.1 | 1 | 229 | |||
| NM_001322293.1 | NP_001309222.1 | 1 | 229 | |||
| GenBank mRNA | AK092633.1 | - | 1 | 188 | ||
| AK294965.1 | - | 1 | 188 | |||
| BC132797.1 | - | 1 | 125 | |||
| BX282552.1 | - | 1 | 202 | |||
| DB221901.1 | - | 1 | 185 | |||
| DB505213.1 | - | 1 | 230 | |||

ZNF391
Genomic Map