siRNA Details
| Gene Symbol: TMEM151B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001137560.1 | NP_001131032.1 | 3 | 3010 | ||
| GenBank mRNA | AK094365.1 | - | 1 | 1011 | ||
| AK124502.1 | - | 1 | 1508 | |||
| L35592.1 | - | 1 | 543 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM151B| Gene Symbol: TMEM151B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001137560.1 | NP_001131032.1 | 3 | 3010 | ||
| GenBank mRNA | AK094365.1 | - | 1 | 1011 | ||
| AK124502.1 | - | 1 | 1508 | |||
| L35592.1 | - | 1 | 543 | |||