siRNA Details
Gene Symbol: PTCHD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017010892.1 | XP_016866381.1 | 2 | 497 | ||
XM_017010893.1 | XP_016866382.1 | 2 | 497 | |||
XM_017010896.1 | XP_016866385.1 | 2 | 497 |
