siRNA Details
| Gene Symbol: EVADR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125857.1 | 2 | 384 | |||
| GenBank mRNA | BU620503.1 | - | 3 | 302 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

EVADR| Gene Symbol: EVADR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125857.1 | 2 | 384 | |||
| GenBank mRNA | BU620503.1 | - | 3 | 302 | ||