siRNA Details
| Gene Symbol: SNHG5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK311192.1 | - | 1 | 426 | ||
| BC009220.2 | - | 1 | 395 | |||
| BC108291.1 | - | 1 | 413 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SNHG5| Gene Symbol: SNHG5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK311192.1 | - | 1 | 426 | ||
| BC009220.2 | - | 1 | 395 | |||
| BC108291.1 | - | 1 | 413 | |||