siRNA Details
| Gene Symbol: ISPD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101426.3 | NP_001094896.1 | 3 | 587 | ||
| XM_006715770.3 | XP_006715833.1 | 3 | 757 | |||
| XM_011515499.2 | XP_011513801.1 | 3 | 732 | |||
| XM_011515500.2 | XP_011513802.1 | 3 | 732 | |||
| XM_011515501.2 | XP_011513803.1 | 3 | 760 | |||
| XM_011515508.2 | XP_011513810.1 | 3 | 1089 | |||
| XM_011515509.2 | XP_011513811.1 | 3 | 1938 | |||
| XM_017012575.1 | XP_016868064.1 | 3 | 732 | |||
| XM_017012576.1 | XP_016868065.1 | 3 | 744 | |||
| XM_017012577.1 | XP_016868066.1 | 2 | 849 | |||
| GenBank mRNA | DT932748.1 | - | 4 | 336 | ||

ISPD
Genomic Map