siRNA Details
| Gene Symbol: HNRNPA2B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002137.3 | NP_002128.1 | 11 | 2523 | ||
| NM_031243.2 | NP_112533.1 | 12 | 2559 | |||
| XM_017012109.1 | XP_016867598.1 | 11 | 1819 | |||
| GenBank mRNA | AK026373.1 | - | 1 | 651 | ||
| BC045724.1 | - | 1 | 385 | |||
| BX537494.1 | - | 9 | 5513 | |||
| BX538099.1 | - | 1 | 1458 | |||

HNRNPA2B1
Genomic Map