siRNA Details
| Gene Symbol: TSPAN33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178562.4 | NP_848657.1 | 8 | 2816 | ||
| GenBank mRNA | AW089489.1 | - | 1 | 118 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TSPAN33| Gene Symbol: TSPAN33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178562.4 | NP_848657.1 | 8 | 2816 | ||
| GenBank mRNA | AW089489.1 | - | 1 | 118 | ||