siRNA Details
| Gene Symbol: SLC37A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011516626.2 | XP_011514928.1 | 1 | 97 | ||
| XM_011516627.2 | XP_011514929.1 | 1 | 96 | |||
| XM_017012712.1 | XP_016868201.1 | 1 | 73 | |||
| XM_017012713.1 | XP_016868202.1 | 1 | 92 | |||
| XM_017012714.1 | XP_016868203.1 | 1 | 113 | |||
| XM_017012715.1 | XP_016868204.1 | 1 | 116 | |||
| XM_017012716.1 | XP_016868205.1 | 1 | 120 | |||
| XM_017012717.1 | XP_016868206.1 | 1 | 92 | |||
| XM_017012718.1 | XP_016868207.1 | 1 | 91 | |||
| GenBank mRNA | AK131347.1 | - | 1 | 201 | ||

SLC37A3
Genomic Map