siRNA Details
| Gene Symbol: LOC101927506 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017014131.1 | XP_016869620.1 | 2, 3 | 2106 | ||
| XM_017014132.1 | XP_016869621.1 | 2, 3 | 2106 | |||
| XM_017014133.1 | XP_016869622.1 | 2, 3 | 2106 | |||

LOC101927506
Genomic Map