siRNA Details
| Gene Symbol: LOC101929709 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BQ007151.1 | - | 1 | 397 | ||
| BX538329.1 | - | 1 | 3418 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929709| Gene Symbol: LOC101929709 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BQ007151.1 | - | 1 | 397 | ||
| BX538329.1 | - | 1 | 3418 | |||