siRNA Details
| Gene Symbol: ZNF252P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_023392.1 | 2 | 99 | |||
| GenBank mRNA | AF086559.1 | - | 2 | 190 | ||
| AK021796.1 | - | 2 | 50 | |||
| AK294723.1 | - | 2 | 57 | |||
| AK296398.1 | - | 2 | 95 | |||
| BC065734.1 | - | 2 | 78 | |||
| DA489661.1 | - | 2 | 99 | |||
| DB233439.1 | - | 2 | 96 | |||

ZNF252P
Genomic Map