siRNA Details
| Gene Symbol: KIAA2026 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017969.2 | NP_001017969.2 | 3 | 1753 | ||
| XM_005251385.2 | XP_005251442.1 | 3 | 2229 | |||
| XM_011517759.2 | XP_011516061.1 | 3 | 2229 | |||
| XM_011517760.2 | XP_011516062.1 | 3 | 2229 | |||
| XM_017014331.1 | XP_016869820.1 | 3 | 2229 | |||
| XM_017014335.1 | XP_016869824.1 | 3 | 2229 | |||
| XM_017014336.1 | XP_016869825.1 | 3 | 2229 | |||

KIAA2026
Genomic Map