siRNA Details
| Gene Symbol: LINC01239 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038977.1 | 5 | 1038 | |||
| GenBank mRNA | AK092601.1 | - | 5 | 1038 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01239| Gene Symbol: LINC01239 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038977.1 | 5 | 1038 | |||
| GenBank mRNA | AK092601.1 | - | 5 | 1038 | ||