siRNA Details
| Gene Symbol: GLIDR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126046.1 | 2 | 720 | |||
| GenBank mRNA | BC019880.1 | - | 2 | 231 | ||
| Gene Symbol: LOC728903 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK093722.1 | - | 1 | 1825 | ||

GLIDR
Genomic Map