siRNA Details
| Gene Symbol: SPANXN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009613.2 | NP_001009613.1 | 2 | 218 | ||
| XM_017029543.1 | XP_016885032.1 | 2 | 208 | |||
| XM_017029544.1 | XP_016885033.1 | 2 | 205 | |||
| GenBank mRNA | BC130503.1 | - | 2 | 218 | 40146534 | |
| BC130505.1 | - | 2 | 218 | 40146536 | ||
| DQ336126.1 | - | 2 | 121 | |||
| DQ336127.1 | - | 2 | 121 | |||

SPANXN4
Genomic Map