siRNA Details
| Gene Symbol: TTTY6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001527.2 | 1 | 93 | |||
| Gene Symbol: TTTY6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002175.3 | 1 | 71 | |||
| GenBank mRNA | AA884105.1 | - | 3 | 399 | ||

TTTY6
Genomic Map