siRNA Details
Gene Symbol: LOC100130698 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136612.1 | 6 | 699 | |||
GenBank mRNA | AK125824.1 | - | 1 | 2930 | ||
AK128140.1 | - | 1 | 1415 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100130698 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136612.1 | 6 | 699 | |||
GenBank mRNA | AK125824.1 | - | 1 | 2930 | ||
AK128140.1 | - | 1 | 1415 |