siRNA Details
| Gene Symbol: MUC19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173600.2 | NP_775871.2 | 4185 | |||
| GenBank mRNA | HM801842.1 | - | 4185 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MUC19| Gene Symbol: MUC19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173600.2 | NP_775871.2 | 4185 | |||
| GenBank mRNA | HM801842.1 | - | 4185 | |||