siRNA Details
| Gene Symbol: TARM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135686.2 | NP_001129158.2 | 5 | 740 | ||
| XM_005278278.3 | XP_005278335.1 | 5 | 1009 | |||
| GenBank mRNA | AK301730.1 | - | 5 | 1025 | ||
| DQ479398.1 | - | 5 | 740 | |||

TARM1
Genomic Map