siRNA Details
| Gene Symbol: LOC646743 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY726562.1 | - | 2 | 194 | ||
| Gene Symbol: TISP43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY713301.1 | - | 2 | 169 | ||

LOC646743
Genomic Map