siRNA Details
| Gene Symbol: LOC105374620 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136207.1 | 2 | 147 | |||
| GenBank mRNA | HY017505.1 | - | 2 | 147 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105374620| Gene Symbol: LOC105374620 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136207.1 | 2 | 147 | |||
| GenBank mRNA | HY017505.1 | - | 2 | 147 | ||