siRNA Details
Gene Symbol: YWHAEP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029404.1 | 1 | 315 | |||
GenBank mRNA | DB053774.1 | - | 1 | 274 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: YWHAEP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029404.1 | 1 | 315 | |||
GenBank mRNA | DB053774.1 | - | 1 | 274 |