siRNA Details
| Gene Symbol: PNMA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001103150.1 | NP_001096620.1 | 2 | 2568 | ||
| NM_001103151.1 | NP_001096621.1 | 3 | 2468 | |||
| NM_001184924.1 | NP_001171853.1 | 4 | 2553 | |||
| NM_052926.2 | NP_443158.1 | 2 | 2554 | |||
| XM_017029252.1 | XP_016884741.1 | 4 | 2870 | |||
| XM_017029253.1 | XP_016884742.1 | 3 | 2532 | |||
| GenBank mRNA | AB067521.1 | - | 2 | 2551 | ||
| AY121806.1 | - | 2 | 2551 | |||
| BC101111.1 | - | 2 | 2412 | 40011824 | ||
| BC101112.2 | - | 2 | 2398 | 40011826 | ||
| BC101113.1 | - | 2 | 2398 | 40011827 | ||

PNMA5
Genomic Map