siRNA Details
| Gene Symbol: H2AFB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080720.1 | NP_542451.1 | 1 | 285 | ||
| GenBank mRNA | AF254576.1 | - | 1 | 285 | ||
| BC101409.1 | - | 1 | 277 | 40021547 | ||
| BC101415.1 | - | 1 | 277 | 40021541 | ||
| BC101417.1 | - | 1 | 277 | 40021543 | ||
| BC101418.1 | - | 1 | 277 | 40021544 | ||
| BC130510.1 | - | 1 | 353 | 40146541 | ||
| BC130512.1 | - | 1 | 353 | 40146543 | ||
| BC134365.1 | - | 1 | 280 | 40021542 | ||
| Gene Symbol: H2AFB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017990.1 | NP_001017990.1 | 1 | 285 | ||
| GenBank mRNA | BC128034.1 | - | 1 | 277 | 40021540 | |
| CR542230.1 | - | 1 | 277 | |||
| Gene Symbol: H2AFB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017991.2 | NP_001017991.1 | 1 | 358 | ||
| GenBank mRNA | HY000925.1 | - | 1 | 359 | ||

H2AFB3
Genomic Map