siRNA Details
| Gene Symbol: LOC101927021 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110065.1 | 4 | 2369 | |||
| NR_110066.1 | 3 | 2051 | ||||
| NR_110067.1 | 4 | 2307 | ||||
| NR_110068.1 | 2 | 2007 | ||||
| NR_110069.1 | 3 | 2199 | ||||
| NR_110070.1 | 5 | 2158 | ||||
| GenBank mRNA | AK090593.1 | - | 3 | 1933 | ||

LOC101927021
Genomic Map