siRNA Details
| Gene Symbol: HNRNPA2B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002137.3 | NP_002128.1 | 11 | 3178 | ||
| NM_031243.2 | NP_112533.1 | 12 | 3214 | |||
| XM_005249729.1 | XP_005249786.1 | 12 | 1318 | |||
| XM_017012109.1 | XP_016867598.1 | 11 | 2474 | |||
| XM_017012110.1 | XP_016867599.1 | 11 | 1284 | |||
| GenBank mRNA | AK026373.1 | - | 1 | 1306 | ||
| BC045724.1 | - | 2 | 508 | |||
| BX537494.1 | - | 9 | 6168 | |||
| BX538099.1 | - | 2 | 1581 | |||

HNRNPA2B1
Genomic Map